ADV509448065 AChemBlock

Methyl 3-iodo-1H-pyrrolo[2,3-b]pyridine-4-carboxylate

  • MDL number MFCD12962693
  • Cas Number : 1190313-88-2
  • Molecular Weight : 302.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-iodo-1h-pyrrolo-2-3-b-pyridine-4-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14200
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart