ADV509448066 AChemBlock

1-(Tetrahydro-2h-pyran-4-yl)-1H-pyrazol-4-amine

  • MDL number MFCD11505007
  • Cas Number : 1190380-49-4
  • Molecular Weight : 167.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-tetrahydro-2h-pyran-4-yl-1h-pyrazol-4-amine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14201
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart