ADV509448067 AChemBlock

(1,3-Oxazol-5-yl)methanamine hydrochloride

  • MDL number MFCD06804578
  • Cas Number : 1196156-45-2
  • Molecular Weight : 134.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-3-oxazol-5-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14202
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart