ADV509448071 AChemBlock

1-{2-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]ethyl}pyrrolidin-2-one

  • MDL number MFCD21337760
  • Cas Number : 1342834-87-0
  • Molecular Weight : 305.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-2-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-ethyl-pyrrolidin-2-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14206
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart