ADV509448073 AChemBlock

4-Bromo-3,5-difluoro-2-iodoaniline

  • MDL number MFCD28133736
  • Cas Number : 1467060-36-1
  • Molecular Weight : 333.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-3-5-difluoro-2-iodoaniline.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14208
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart