ADV509448076 AChemBlock

methyl (2S,4S)-4-((tert-butoxycarbonyl)amino)pyrrolidine-2-carboxylate

  • MDL number MFCD08704523
  • Cas Number : 168263-82-9
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/l-proline-4-1-1-dimethylethoxy-carbonyl-amino-methyl-ester-4s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14211
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart