ADV509448081 AChemBlock

(S)-1-isopropylazetidine-2-carboxylic acid

  • MDL number MFCD11053659
  • Cas Number : 255882-98-5
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-azetidinecarboxylic-acid-1-1-methylethyl-2s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14216
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart