ADV509448091 AChemBlock

(2S,4R)-4-methoxy-1-methylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD29767916
  • Cas Number : 1860012-51-6
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/l-proline-4-methoxy-1-methyl-trans-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14227
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart