ADV509448099 AChemBlock

(3S)-3-Aminopyrrolidin-2-one hydrochloride

  • MDL number MFCD08273939
  • Cas Number : 56440-28-9
  • Molecular Weight : 136.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3s-3-aminopyrrolidin-2-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14236
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart