ADV509448110 AChemBlock

8-Chloro-2-methoxy-1,5-naphthyridine

  • MDL number MFCD09027618
  • Cas Number : 249889-68-7
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/8-chloro-2-methoxy-1-5-naphthyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14247
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart