ADV509448117 AChemBlock

[(2S)-4-Benzylpiperazin-2-yl]methanol dihydrochloride

  • MDL number MFCD22392720
  • Cas Number : 1860012-50-5
  • Molecular Weight : 279.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2s-4-benzylpiperazin-2-yl-methanol-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14254
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart