ADV509448121 AChemBlock

3-Chloro-4-nitro-1H-pyrazole

  • MDL number MFCD16619868
  • Cas Number : 35852-75-6
  • Molecular Weight : 147.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-4-nitro-1h-pyrazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14258
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart