ADV509448129 AChemBlock

(2R,4R)-4-methoxy-1-methylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD30146456
  • Cas Number : 1215385-33-3
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/d-proline-4-methoxy-1-methyl-4r-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14266
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart