ADV509448136 AChemBlock

(5-Fluoropyrimidin-2-yl)methanamine hydrochloride

  • MDL number MFCD20230580
  • Cas Number : 1314394-77-8
  • Molecular Weight : 163.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-fluoropyrimidin-2-yl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14273
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart