ADV509448144 AChemBlock

1-(tert-butyl) 3-methyl 3-aminoazetidine-1,3-dicarboxylate

  • MDL number MFCD28631502
  • Cas Number : 1782647-31-7
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-3-azetidinedicarboxylic-acid-3-amino-1-1-1-dimethylethyl-3-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14281
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart