ADV509448151 AChemBlock

tert-butyl (2S,4S)-4-hydroxy-2-methylpyrrolidine-1-carboxylate

  • MDL number MFCD20278358
  • Cas Number : 477293-60-0
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-pyrrolidinecarboxylic-acid-4-hydroxy-2-methyl-1-1-dimethylethyl-ester-2s-4s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14288
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart