ADV509448158 AChemBlock

5-Fluoro-6-methylpyridine-2-carbaldehyde

  • MDL number MFCD08277273
  • Cas Number : 884495-34-5
  • Molecular Weight : 139.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-fluoro-6-methylpyridine-2-carbaldehyde.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14295
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart