ADV509448163 AChemBlock

Ethyl 2-(5-{[(tert-butoxy)carbonyl]amino}pyridin-2-yl)acetate

  • MDL number MFCD23701733
  • Cas Number : 921940-82-1
  • Molecular Weight : 280.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-5-tert-butoxy-carbonyl-amino-pyridin-2-yl-acetate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14300
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart