ADV509448171 AChemBlock

2-Oxo-1H,2H,3H-pyrrolo[2,3-b]pyridine-5-carboxylic acid

  • MDL number MFCD18250900
  • Cas Number : 1260665-66-4
  • Molecular Weight : 178.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-oxo-1h-2h-3h-pyrrolo-2-3-b-pyridine-5-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14308
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart