ADV509448174 AChemBlock

1-(Pyrazin-2-yl)cyclopropane-1-carboxylic acid

  • MDL number MFCD17926215
  • Cas Number : 1159734-52-7
  • Molecular Weight : 164.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-pyrazin-2-yl-cyclopropane-1-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14311
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart