ADV509448181 AChemBlock

3-Amino-1-[(tert-butoxy)carbonyl]azetidine-3-carboxylic acid

  • MDL number MFCD18375110
  • Cas Number : 1262412-13-4
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-amino-1-tert-butoxy-carbonyl-azetidine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14318
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart