ADV509448183 AChemBlock

2-Chloro-5H,6H,7H,8H-pyrido[4,3-d]pyrimidine hydrochloride

  • MDL number MFCD18434487
  • Cas Number : 1314723-39-1
  • Molecular Weight : 206.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5h-6h-7h-8h-pyrido-4-3-d-pyrimidine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14320
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart