ADV509448204 AChemBlock

Tert-butyl (2R,3S)-2-cyano-3-hydroxypyrrolidine-1-carboxylate

  • MDL number MFCD29918581
  • Cas Number : 1946010-83-8
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-3s-2-cyano-3-hydroxypyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14342
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart