ADV509448212 AChemBlock

1H-Indole-5-sulfonyl chloride

  • MDL number MFCD19200539
  • Cas Number : 1094209-33-2
  • Molecular Weight : 215.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1h-indole-5-sulfonyl-chloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14350
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart