ADV509448223 AChemBlock

3-Benzyl-3-azabicyclo[3.3.1]nonan-9-one

  • MDL number MFCD10666933
  • Cas Number : 81879-64-3
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-benzyl-3-azabicyclo-3-3-1-nonan-9-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14361
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart