ADV509448232 AChemBlock

methyl (S)-1-(2-fluoroethyl)azetidine-2-carboxylate

  • MDL number MFCD29918539
  • Cas Number : 1860033-52-8
  • Molecular Weight : 161.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-azetidinecarboxylic-acid-1-2-fluoroethyl-methyl-ester-2s.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14370
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart