ADV509448238 AChemBlock

(1-Fluorocyclobutyl)methanamine hydrochloride

  • MDL number MFCD27922430
  • Cas Number : 1462885-81-9
  • Molecular Weight : 139.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/1-fluorocyclobutyl-methanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14376
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart