ADV509448248 AChemBlock

5-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-tosyl-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD16039195
  • Cas Number : 866546-11-4
  • Molecular Weight : 432.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1-tosyl-1h-pyrrolo-2-3-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14386
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart