ADV509448250 AChemBlock

5-Chloro-3H,4H-pyrrolo[2,1-f][1,2,4]triazin-4-one

  • MDL number MFCD23105893
  • Cas Number : 888720-60-3
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-3h-4h-pyrrolo-2-1-f-1-2-4-triazin-4-one.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14388
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart