ADV509448251 AChemBlock

Methyl 1,2,3,4-tetrahydroisoquinoline-5-carboxylate hydrochloride

  • MDL number MFCD03094738
  • Cas Number : 1035700-06-1
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-2-3-4-tetrahydroisoquinoline-5-carboxylate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14389
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart