ADV509448267 AChemBlock

7H-Pyrrolo[2,3-d]pyrimidin-4-ylmethanamine hydrochloride

  • MDL number MFCD19689066
  • Cas Number : 935466-96-9
  • Molecular Weight : 184.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/7h-pyrrolo-2-3-d-pyrimidin-4-ylmethanamine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14405
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart