ADV509448270 AChemBlock

2-(Trifluoromethyl)cyclopropan-1-amine hydrochloride

  • MDL number MFCD13806389
  • Cas Number : 1172072-62-6
  • Molecular Weight : 161.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-trifluoromethyl-cyclopropan-1-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14408
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart