ADV509448281 AChemBlock

tert-Butyl 5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylate

  • MDL number MFCD17214286
  • Cas Number : 1256359-17-7
  • Molecular Weight : 294.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14420
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart