ADV509448282 AChemBlock

6-Iodo-2-methyl-1H-indole

  • MDL number MFCD18380835
  • Cas Number : 1260383-44-5
  • Molecular Weight : 257.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-iodo-2-methyl-1h-indole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14421
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart