ADV509448293 AChemBlock

5-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyridine

  • MDL number MFCD29060202
  • Cas Number : 1784380-03-5
  • Molecular Weight : 232.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-chloro-1h-pyrazolo-3-4-b-pyridine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14432
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart