ADV509448295 AChemBlock

(2S,4S)-1,4-Dimethylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD30146464
  • Cas Number : 6734-42-5
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2s-4s-1-4-dimethylpyrrolidine-2-carboxylic-acid-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14434
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart