ADV509448299 AChemBlock

5-Methyl-5H-pyrrolo[2,3-b]pyrazine-7-carboxylic acid methyl ester

  • MDL number MFCD29918561
  • Cas Number : 1860028-29-0
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-5h-pyrrolo-2-3-b-pyrazine-7-carboxylic-acid-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14438
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart