ADV509448307 AChemBlock

(4-Fluoropyridin-3-yl)methanol hydrochloride

  • MDL number MFCD30146466
  • Cas Number : 1946021-34-6
  • Molecular Weight : 163.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-fluoropyridin-3-yl-methanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14446
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart