ADV509448311 AChemBlock

3-(Trifluoromethyl)-5H,6H,7H,8H-imidazo[1,5-a]pyrazine

  • MDL number MFCD09954909
  • Cas Number : 959238-29-0
  • Molecular Weight : 191.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/3-trifluoromethyl-5h-6h-7h-8h-imidazo-1-5-a-pyrazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14450
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart