ADV509448312 AChemBlock

(2S,4S)-Tert-butyl 2-(difluoromethyl)-4-hydroxypyrrolidine-1-carboxylate

  • MDL number MFCD29991175
  • Cas Number : 1207852-93-4
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2s-4s-tert-butyl-2-difluoromethyl-4-hydroxypyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14451
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart