ADV509448313 AChemBlock

(r)-(3-Pyrrolidin-2-yl-propyl)-carbamic acid tert-butyl ester

  • MDL number MFCD06656824
  • Cas Number : 1217838-01-1
  • Molecular Weight : 228.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/r-3-pyrrolidin-2-yl-propyl-carbamic-acid-tert-butyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14452
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart