ADV509448322 AChemBlock

Imidazo[1,5-a]pyridin-1-ylmethanol

  • MDL number MFCD19105076
  • Cas Number : 112230-23-6
  • Molecular Weight : 148.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/imidazo-1-5-a-pyridin-1-ylmethanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14461
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart