ADV509448328 AChemBlock

5-tert-Butyl 3-ethyl 4H,5H,6H,7H-thieno[3,2-c]pyridine-3,5-dicarboxylate

  • MDL number MFCD23105853
  • Cas Number : 1363381-39-8
  • Molecular Weight : 311.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-tert-butyl-3-ethyl-4h-5h-6h-7h-thieno-3-2-c-pyridine-3-5-dicarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14467
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart