ADV509448339 AChemBlock

5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-benzoxazole

  • MDL number MFCD09027764
  • Cas Number : 837392-66-2
  • Molecular Weight : 245.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/5-tetramethyl-1-3-2-dioxaborolan-2-yl-1-2-benzoxazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14478
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart