ADV509448342 AChemBlock

4-(Aminomethyl)-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one dihydrochloride

  • MDL number MFCD29060190
  • Cas Number : 1818847-61-8
  • Molecular Weight : 236.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/4-aminomethyl-1h-2h-3h-pyrrolo-2-3-b-pyridin-2-one-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14481
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart