ADV509448343 AChemBlock

N-Cyclopropyl-1H-indole-5-sulfonamide

  • MDL number MFCD30146471
  • Cas Number : 1860028-27-8
  • Molecular Weight : 236.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/n-cyclopropyl-1h-indole-5-sulfonamide.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14482
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart