ADV509448348 AChemBlock

2-Aza-bicyclo[2.2.1]heptane-2,6-dicarboxylic acid 2-tert-butyl ester

  • MDL number MFCD30146472
  • Cas Number : 1860028-20-1
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-aza-bicyclo-2-2-1-heptane-2-6-dicarboxylic-acid-2-tert-butyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14487
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart