ADV509448353 AChemBlock

2-Azabicyclo[2.2.1]heptane-6-carboxylic acid hydrochloride

  • MDL number MFCD30146474
  • Cas Number : 1949816-37-8
  • Molecular Weight : 177.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2-azabicyclo-2-2-1-heptane-6-carboxylic-acid-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14492
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart