ADV509448355 AChemBlock

6-[(tert-Butoxy)carbonyl]-5,6,7,8-tetrahydro-1,6-naphthyridine-3-carboxylic acid

  • MDL number MFCD19687760
  • Cas Number : 1341036-28-9
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/6-tert-butoxy-carbonyl-5-6-7-8-tetrahydro-1-6-naphthyridine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14494
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart