ADV509448370 AChemBlock

(2S,5R)-1,5-Dimethylpyrrolidine-2-carboxylic acid

  • MDL number MFCD19230076
  • Cas Number : 1932347-35-7
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://www.achemblock.com/2s-5r-1-5-dimethylpyrrolidine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L14511
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart